Assay Details
| Gene Symbol: Rbm48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654553.1 | XP_007652743.1 | 2 - 3 | 348 | 78 | |
| XM_007654556.1 | XP_007652746.1 | 2 - 3 | 348 | 78 | ||
| XM_007654559.1 | XP_007652749.1 | 2 - 3 | 348 | 78 | ||
| GenBank mRNA | JI863837.1 | - | 2 - 3 | 342 | 78 | |
More Information
Set Membership: |
|

Rbm48
Genomic Map