Assay Details
| Gene Symbol: Cd40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500525.1 | XP_003500573.1 | 4 - 5 | 404 | 61 | |
| XM_003500526.1 | XP_003500574.1 | 4 - 5 | 404 | 61 | ||
More Information
Set Membership: |
|

Cd40
Genomic Map