Assay Details
| Gene Symbol: Trappc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500861.2 | XP_003500909.1 | 1 - 2 | 181 | 64 | |
| GenBank mRNA | JI891856.1 | - | 1 - 2 | 160 | 64 | |
| JP047334.1 | - | 1 - 2 | 160 | 64 | ||
More Information
Set Membership: |
|

Trappc3
Genomic Map