Assay Details
Gene Symbol: Rpl18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007637893.1 | XP_007636083.1 | 4 - 5 | 481 | 55 | |
GenBank mRNA | JI877663.1 | - | 4 - 5 | 478 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
