Assay Details
| Gene Symbol: Fam206a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500753.2 | XP_003500801.1 | 5 - 6 | 539 | 71 | |
| GenBank mRNA | JI886202.1 | - | 5 - 6 | 490 | 71 | |
| JP053643.1 | - | 5 - 6 | 490 | 71 | ||
More Information
Set Membership: |
|

Fam206a
Genomic Map