Assay Details
Gene Symbol: Mbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511839.2 | XP_003511887.1 | 3 - 4 | 503 | 60 | |
GenBank mRNA | JI867833.1 | - | 1 - 2 | 232 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
