Assay Details
| Gene Symbol: Zdhhc11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513952.1 | XP_003514000.1 | 4 - 5 | 609 | 80 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc11b| Gene Symbol: Zdhhc11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513952.1 | XP_003514000.1 | 4 - 5 | 609 | 80 | |