Assay Details
| Gene Symbol: Dsel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508460.2 | XP_003508508.1 | 2 | 4009 | 104 | |
| XM_007649081.1 | XP_007647271.1 | 1 | 3723 | 104 | ||
More Information
Set Membership: |
|

Dsel
Genomic Map