Assay Details
Gene Symbol: LOC103160089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646314.1 | XP_007644504.1 | 2 - 3 | 465 | 92 | |
GenBank mRNA | JP043705.1 | - | 1 - 2 | 193 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
