Assay Details
Gene Symbol: Plcxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643896.1 | XP_007642086.1 | 1 - 2 | 753 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plcxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643896.1 | XP_007642086.1 | 1 - 2 | 753 | 61 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |