Assay Details
Gene Symbol: Rbm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515055.2 | XP_003515103.1 | 1 - 2 | 4636 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
