Assay Details
Gene Symbol: LOC100689011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646345.1 | XP_007644535.1 | 6 - 7 | 1266 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
