Assay Details
| Gene Symbol: LOC100769698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648736.1 | XP_007646926.1 | 2 - 3 | 112 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100769698| Gene Symbol: LOC100769698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648736.1 | XP_007646926.1 | 2 - 3 | 112 | 89 | |
Set Membership: |
|