Assay Details
Gene Symbol: Cdkn2aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641426.1 | XP_007639616.1 | 1 - 2 | 81 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
