Assay Details
| Gene Symbol: Ndufb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649734.1 | XP_007647924.1 | 2 - 3 | 163 | 110 | |
| GenBank mRNA | JI862449.1 | - | 1 - 2 | 147 | 110 | |
More Information
Set Membership: |
|

Ndufb1
Genomic Map