Assay Details
Gene Symbol: Chst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641730.1 | XP_007639920.1 | 1 | 621 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
