Assay Details
| Gene Symbol: LOC100767032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511454.2 | XP_003511502.2 | 12 - 13 | 2133 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100767032| Gene Symbol: LOC100767032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511454.2 | XP_003511502.2 | 12 - 13 | 2133 | 59 | |
Set Membership: |
|