Assay Details
| Gene Symbol: Bst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513630.2 | XP_003513678.1 | 1 - 2 | 200 | 66 | |
| XM_007653887.1 | XP_007652077.1 | 1 - 2 | 200 | 66 | ||
More Information
Set Membership: |
|

Bst1
Genomic Map