Assay Details
Gene Symbol: N4bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007628969.1 | XP_007627159.1 | 2 - 3 | 377 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: N4bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007628969.1 | XP_007627159.1 | 2 - 3 | 377 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |