Assay Details
Gene Symbol: Cpeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506782.2 | XP_003506830.1 | 9 - 10 | 1048 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
