Assay Details
Gene Symbol: Ino80e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510111.2 | XP_003510159.1 | 2 - 3 | 279 | 66 | |
XM_007650742.1 | XP_007648932.1 | 2 - 3 | 279 | 66 | ||
XM_007650743.1 | XP_007648933.1 | 2 - 3 | 279 | 66 | ||
GenBank mRNA | JI893720.1 | - | 2 - 3 | 298 | 66 | |
JP040216.1 | - | 2 - 3 | 298 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
