Assay Details
Gene Symbol: Msantd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644551.1 | XP_007642741.1 | 2 - 3 | 594 | 71 | |
GenBank mRNA | JP062339.1 | - | 1 - 2 | 480 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
