Assay Details
| Gene Symbol: Mob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650958.1 | XP_007649148.1 | 5 - 6 | 715 | 61 | |
| GenBank mRNA | JI870208.1 | - | 2 - 3 | 327 | 61 | |
More Information
Set Membership: |
|

Mob2
Genomic Map