Assay Details
Gene Symbol: LOC100768534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645456.1 | XP_007643646.1 | 10 - 11 | 2796 | 80 | |
XM_007645458.1 | XP_007643648.1 | 10 - 11 | 2819 | 80 | ||
GenBank mRNA | JI895711.1 | - | 7 - 8 | 1148 | 80 | |
JP050643.1 | - | 7 - 8 | 1148 | 80 |
