Assay Details
Gene Symbol: LOC100768534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645456.1 | XP_007643646.1 | 12 - 13 | 3040 | 75 | |
XM_007645458.1 | XP_007643648.1 | 12 - 13 | 3063 | 75 | ||
GenBank mRNA | JI895711.1 | - | 9 - 10 | 1392 | 75 | |
JP050643.1 | - | 9 - 10 | 1392 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
