Assay Details
Gene Symbol: LOC100768534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645456.1 | XP_007643646.1 | 2 - 3 | 213 | 61 | |
XM_007645458.1 | XP_007643648.1 | 2 - 3 | 236 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
