Assay Details
Gene Symbol: LOC100768534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645456.1 | XP_007643646.1 | 8 - 9 | 2616 | 74 | |
XM_007645458.1 | XP_007643648.1 | 8 - 9 | 2639 | 74 | ||
GenBank mRNA | JI895711.1 | - | 5 - 6 | 968 | 74 | |
JP050643.1 | - | 5 - 6 | 968 | 74 |
