Assay Details
Gene Symbol: LOC100768534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645456.1 | XP_007643646.1 | 9 - 10 | 2707 | 66 | |
XM_007645458.1 | XP_007643648.1 | 9 - 10 | 2730 | 66 | ||
GenBank mRNA | JI895711.1 | - | 6 - 7 | 1059 | 66 | |
JP050643.1 | - | 6 - 7 | 1059 | 66 |

More Information
Set Membership: |
![]() ![]() |
