Assay Details
Gene Symbol: Sp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645696.1 | XP_007643886.1 | 1 - 2 | 88 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
