Assay Details
| Gene Symbol: Lclat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496566.2 | XP_003496614.1 | 5 - 6 | 730 | 65 | |
| GenBank mRNA | JI868866.1 | - | 4 - 5 | 615 | 65 | |
| JP050971.1 | - | 4 - 5 | 615 | 65 | ||
More Information
Set Membership: |
|

Lclat1
Genomic Map