Assay Details
| Gene Symbol: Fut10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497838.2 | XP_003497886.1 | 3 - 4 | 757 | 79 | |
| XM_007618452.1 | XP_007616642.1 | 3 - 4 | 757 | 79 | ||
More Information
Set Membership: |
|

Fut10
Genomic Map