Assay Details
| Gene Symbol: Maff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512999.2 | XP_003513047.2 | 2 - 3 | 530 | 90 | |
| XM_007653384.1 | XP_007651574.1 | 2 - 3 | 220 | 90 | ||
| XM_007653385.1 | XP_007651575.1 | 2 - 3 | 335 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007653385.1 transcripts.
Set Membership: |
|

Maff
Genomic Map