Assay Details
Gene Symbol: Fabp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510074.2 | XP_003510122.1 | 2 - 3 | 346 | 58 | |
GenBank mRNA | JI884147.1 | - | 2 - 3 | 483 | 58 | |
JP054072.1 | - | 2 - 3 | 482 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
