Assay Details
Gene Symbol: Arl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651315.1 | XP_007649505.1 | 4 - 5 | 533 | 65 | |
GenBank mRNA | JI870362.1 | - | 4 - 5 | 464 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
