Assay Details
Gene Symbol: Ptcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650320.1 | XP_007648510.1 | 4 - 5 | 1000 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
