Assay Details
Gene Symbol: Hoxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642618.1 | XP_007640808.1 | 3 - 4 | 300 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
