Assay Details
Gene Symbol: Hcfc1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508712.2 | XP_003508760.1 | 1 - 2 | 240 | 92 | |
GenBank mRNA | JP059050.1 | - | 1 - 2 | 284 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
