Assay Details
| Gene Symbol: Rps15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514958.2 | XP_003515006.1 | 2 - 3 | 171 | 92 | |
| XM_007654980.1 | XP_007653170.1 | 2 - 3 | 419 | 92 | ||
| GenBank mRNA | JI867097.1 | - | 2 - 3 | 192 | 92 | |
| JP064747.1 | - | 2 - 3 | 192 | 92 | ||
More Information
Set Membership: |
|

Rps15a
Genomic Map