Assay Details
Gene Symbol: Cidec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653773.1 | XP_007651963.1 | 2 - 3 | 230 | 81 | |
GenBank mRNA | JI866430.1 | - | 1 - 2 | 187 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
