Assay Details
Gene Symbol: Chst13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007616172.1 | XP_007614362.1 | 1 - 2 | 105 | 106 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
