Assay Details
Gene Symbol: Trnau1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498376.2 | XP_003498424.1 | 7 - 8 | 696 | 61 | |
XM_007626854.1 | XP_007625044.1 | 6 - 7 | 576 | 61 | ||
XM_007626860.1 | XP_007625050.1 | 6 - 7 | 564 | 61 | ||
XM_007626867.1 | XP_007625057.1 | 5 - 6 | 543 | 61 | ||
GenBank mRNA | JI868440.1 | - | 1 - 2 | 160 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
