Assay Details
Gene Symbol: Elmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511936.2 | XP_003511984.1 | 5 - 6 | 639 | 72 | |
GenBank mRNA | JI877639.1 | - | 3 - 4 | 364 | 72 | |
JP063723.1 | - | 3 - 4 | 364 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
