Assay Details
Gene Symbol: Znf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007607793.1 | XP_007605983.1 | 1 - 2 | 768 | 65 | |
XM_007607794.1 | XP_007605984.1 | 2 - 3 | 732 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
