Assay Details
Gene Symbol: Eif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649676.1 | XP_007647866.1 | 1 - 2 | 296 | 81 | |
GenBank mRNA | JI892363.1 | - | 1 - 2 | 296 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
