Assay Details
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501887.2 | XP_003501935.1 | 1 - 2 | 193 | 60 | |
GenBank mRNA | JI887633.1 | - | 1 - 2 | 195 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
