Assay Details
| Gene Symbol: Dut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506245.2 | XP_003506293.2 | 1 - 2 | 142 | 61 | |
| XM_007646749.1 | XP_007644939.1 | 1 - 2 | 145 | 61 | ||
More Information
Set Membership: |
|

Dut
Genomic Map