Assay Details
Gene Symbol: Obfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515103.2 | XP_003515151.1 | 6 - 7 | 928 | 62 | |
XM_007655107.1 | XP_007653297.1 | 6 - 7 | 787 | 62 | ||
XM_007655108.1 | XP_007653298.1 | 6 - 7 | 804 | 62 | ||
XM_007655109.1 | XP_007653299.1 | 5 - 6 | 760 | 62 | ||
GenBank mRNA | JI863915.1 | - | 1 - 2 | 122 | 62 | |
JP043113.1 | - | 1 - 2 | 122 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
