Assay Details
Gene Symbol: Obfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515103.2 | XP_003515151.1 | 7 - 8 | 1104 | 63 | |
XM_007655107.1 | XP_007653297.1 | 7 - 8 | 963 | 63 | ||
XM_007655108.1 | XP_007653298.1 | 7 - 8 | 980 | 63 | ||
XM_007655109.1 | XP_007653299.1 | 6 - 7 | 936 | 63 | ||
GenBank mRNA | JI863915.1 | - | 2 - 3 | 298 | 63 | |
JP043113.1 | - | 2 - 3 | 298 | 63 |

More Information
Set Membership: |
![]() ![]() |
