Assay Details
| Gene Symbol: Bricd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642208.1 | XP_007640398.1 | 1 - 2 | 2357 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bricd5| Gene Symbol: Bricd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642208.1 | XP_007640398.1 | 1 - 2 | 2357 | 58 | |
Set Membership: |
|