Assay Details
| Gene Symbol: Slc25a44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502916.2 | XP_003502964.1 | 3 - 4 | 1252 | 67 | |
| XM_007643535.1 | XP_007641725.1 | 3 - 4 | 1250 | 67 | ||
More Information
Set Membership: |
|

Slc25a44
Genomic Map